Learn More
Medchemexpress LLC Idazoxan (hydrochloride) | 79944-56-2 | 99.3% | C11H13ClN2O2 | 25 MG
SDP
Catalog No. 5000356620
Encompass_Preferred
Idazoxan hydrochloride is an α2-adrenoceptor and imidazoline receptor antagonist that competitively blocks the hypotensive effect of imidazoline-like agents. It also shows potential in improving motor symptoms associated with Parkinson's disease, L-DOPA-induced dyskinesias, and experimental Parkinsonism.
  • Acts as an alpha-2 adrenoceptor antagonist.
  • Functions as an imidazoline receptor antagonist.
  • Antagonizes centrally induced hypotensive effects.
  • May improve motor symptoms in Parkinson's disease.
  • May alleviate L-DOPA-induced dyskinesias.
  • Applicable to experimental Parkinsonism research.
Catalog No. 50-003-56620 Supplier Medchemexpress LLC Supplier No. 25MGHY14561A
Idazoxan hydrochloride is an α2-adrenoceptor and imidazoline receptor antagonist that competitively blocks the hypotensive effect of imidazoline-like agents. It also shows potential in improving motor symptoms associated with Parkinson's disease, L-DOPA-induced dyskinesias, and experimental Parkinsonism.
  • Acts as an alpha-2 adrenoceptor antagonist.
  • Functions as an imidazoline receptor antagonist.
  • Antagonizes centrally induced hypotensive effects.
  • May improve motor symptoms in Parkinson's disease.
  • May alleviate L-DOPA-induced dyskinesias.
  • Applicable to experimental Parkinsonism research.