Learn More
Medchemexpress LLC Idazoxan hydrochloride | 79944-56-2 | MFCD00069293 | 99.32% | C11H13ClN2O2 | 1 ML
SDP
Catalog No. 5000356371
Encompass_Preferred
Idazoxan hydrochloride is a compound that acts as an α2-adrenoceptor antagonist and an imidazoline receptors antagonist. It is known for its ability to competitively antagonize the centrally induced hypotensive effect of imidazoline-like agents. Research indicates its potential to improve motor symptoms associated with Parkinson's disease, L-DOPA-induced dyskinesias, and experimental Parkinsonism.
  • Acts as an α2-adrenoceptor antagonist
  • Functions as an imidazoline receptors antagonist
  • Competitively antagonizes centrally induced hypotensive effects
  • Potential to improve motor symptoms in Parkinson's disease
  • Potential to improve L-DOPA-induced dyskinesias
  • Potential to improve experimental Parkinsonism
  • Reverses haloperidol-induced catalepsy in rat models
Catalog No. 50-003-56371 Supplier Medchemexpress LLC Supplier No. HY14561A10MM/1ML
Idazoxan hydrochloride is a compound that acts as an α2-adrenoceptor antagonist and an imidazoline receptors antagonist. It is known for its ability to competitively antagonize the centrally induced hypotensive effect of imidazoline-like agents. Research indicates its potential to improve motor symptoms associated with Parkinson's disease, L-DOPA-induced dyskinesias, and experimental Parkinsonism.
  • Acts as an α2-adrenoceptor antagonist
  • Functions as an imidazoline receptors antagonist
  • Competitively antagonizes centrally induced hypotensive effects
  • Potential to improve motor symptoms in Parkinson's disease
  • Potential to improve L-DOPA-induced dyskinesias
  • Potential to improve experimental Parkinsonism
  • Reverses haloperidol-induced catalepsy in rat models