Learn More
Abcam Idazo x an hydrochloride, alpha2 Adrenoceptor antagonist and I2 ligand, 10MG
Catalog No. 502706870
Encompass_Preferred
MW 240.68 Da, Purity >99%. α2 Adrenoceptor antagonist and I2 ligand. Improves motor symptoms and decreases dyskinesia in Parkinson's disease models. Also induces apoptosis. Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6870 Supplier Abcam Supplier No. AB14113010MG
MW 240.68 Da, Purity >99%. α2 Adrenoceptor antagonist and I2 ligand. Improves motor symptoms and decreases dyskinesia in Parkinson's disease models. Also induces apoptosis. Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement