Learn More
3M™ IDBLUE™ Ultra-Mobile HF RFID Bluetooth™ Reader
Catalog No. 19071130 Shop All 3M Products
Change view
Click to view available options
mobile reader
Catalog No.
19-071-130 mobile reader
1 options
Catalog No. 19-071-130 Supplier 3M™ Supplier No. 7100107843
3M™ IDBLUE™ Ultra-Mobile HF RFID Bluetooth™ Reader is compatible with and easily interchanged with many different types of platforms
  • Ergonomically designed pen style shape
  • Simple press-and-release single button user interface
  • Integrated Bluetooth™ 2.1
  • FCC, CE, and IP54 Certified
  • 3M Catalog# IDBLUE-HF
TRUSTED_SUSTAINABILITY
Specifications
Product Type RFID Bluetooth Reader
Model IDBLUE-HF
For Use With (Equipment) Connected Safety