Learn More
Apexbio Technology LLC IDE 1 1160927-48-9 10mg
SDP
Catalog No. 5000091881
Encompass_Preferred
IDE 1 (CAS 1160927-48-9) is a small-molecule compound designed to promote differentiation of stem cells toward the definitive endoderm lineage by activating receptors involved in the TGF- signaling pathway IDE 1 exerts its biological activity primarily through activation of the TGF- signaling pathway In in vitro studies IDE 1 induces definitive endoderm specification from pluripotent stem cells Based on these pharmacological properties IDE 1 holds research potential in in vitro differentiation protocols to generate endoderm-derived cells such as hepatocytes and pancreatic cells and in studies investigating early embryonic lineage commitment and regenerative medicine
Catalog No. 50-000-91881 Supplier Apexbio Technology LLC Supplier No. B756610
IDE 1 (CAS 1160927-48-9) is a small-molecule compound designed to promote differentiation of stem cells toward the definitive endoderm lineage by activating receptors involved in the TGF- signaling pathway IDE 1 exerts its biological activity primarily through activation of the TGF- signaling pathway In in vitro studies IDE 1 induces definitive endoderm specification from pluripotent stem cells Based on these pharmacological properties IDE 1 holds research potential in in vitro differentiation protocols to generate endoderm-derived cells such as hepatocytes and pancreatic cells and in studies investigating early embryonic lineage commitment and regenerative medicine