Learn More
Abcam IDE 2, endoderm formation inducer, 1MG
Catalog No. 502705385
Encompass_Preferred
MW 240.3 Da, Purity >98%. Cell-permeable definitive endoderm formation inducer (EC₅₀ = 223 nM, induction of Sox17 expression in ESCs) similar to IDE 1 (ab145222). Activates TGF-β signaling and downstream Smad2 phosphorylation. Upregulates Nodal expression. Induces gut tube formation in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5385 Supplier Abcam Supplier No. AB1452231MG
Add to Cart
Add to Cart
MW 240.3 Da, Purity >98%. Cell-permeable definitive endoderm formation inducer (EC₅₀ = 223 nM, induction of Sox17 expression in ESCs) similar to IDE 1 (ab145222). Activates TGF-β signaling and downstream Smad2 phosphorylation. Upregulates Nodal expression. Induces gut tube formation in vivo. .
The product is subject to the following: Abcam Restricted Use Statement