Learn More
STEMCELL Technologies IDE2, Size: 5 mg
SDP
Catalog No. 501975936
Encompass
Inducer of definitive endoderm 2 (IDE2) induces differentiation of mouse or human pluripotent stem cells (PSCs) by activating SMAD2 phosphorylation and NODAL expression (Borowiak et al.). At EC₅₀ = 223nM, SOX17 expression was induced in mouse ES cells.DIFFERENTIATION· Induces definitive endoderm from mouse or human ES cells in the absence of Activin A, NODAL, or feeder cells (Borowiak et al.).
Catalog No. 50-197-5936 Supplier STEMCELL Technologies Supplier No. 72524
Inducer of definitive endoderm 2 (IDE2) induces differentiation of mouse or human pluripotent stem cells (PSCs) by activating SMAD2 phosphorylation and NODAL expression (Borowiak et al.). At EC₅₀ = 223nM, SOX17 expression was induced in mouse ES cells.DIFFERENTIATION· Induces definitive endoderm from mouse or human ES cells in the absence of Activin A, NODAL, or feeder cells (Borowiak et al.).