Learn More
Diagenode Inc ChIP-seq Kit, 24 reactions, Transcription factors, non-histone proteins, Mammalian, yeast compatibility, 4 million cells per IP, 30 mg tissue per IP
Catalog No. NC0824731
Encompass
ChIP-seq Kit, 24 reactions, Transcription factors, non-histone proteins, Mammalian, yeast compatibility, 4 million cells per IP, 30 mg tissue per IP
Catalog No. NC0824731 Supplier Diagenode Inc Supplier No. C01010055
ChIP-seq Kit, 24 reactions, Transcription factors, non-histone proteins, Mammalian, yeast compatibility, 4 million cells per IP, 30 mg tissue per IP