Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Ideal Gas Law Kit
GREENER_CHOICESCI_ED

Catalog No. S18901 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S18901 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S18901 Supplier Innovating Science™ Supplier No. IS2528
Add to Cart
Add to Cart
This item is not returnable. View return policy

Explore the gas laws through the reaction of sodium bicarbonate and hydrochloric acid with the Ideal Gas Law Kit. Students will perform the reaction and use the ideal gas law to calculate the amount of carbon dioxide produced, then use stoichiometry to determine their percent yield.

  • Contains enough materials for 15 groups of students
  • Teacher Manual and Student Study Guide Copymasters are included
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: PS1.A, PS1.B
  • Performance Expectations: MS-PS1-2, MS-PS1-4
  • Cross Cutting Concepts: Patterns, Cause and Effect Engineering Practices, Analyzing and Interpreting Data, Developing and Using Models

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Ideal Gas Law Education
Includes 40 g Sodium Bicarbonate (1), 25 mL 1M Hydrochloric Acid (15), String (1), Balloons (15), Teacher’s Manual, and Student Study Guide Copymasters
Class Size 15 Lab Groups
Product Type Chemistry Education Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.