Learn More
Braintree Scientific IDEAL MICRO DRILL
SDP
Catalog No. NC9010016
Encompass
Ideal Micro-Drill Designed for research applications that require surgical burrs and trephines. Comes with five burrs: .006mm, .008mm, .010mm, 1.012mm, 1.016mm. Rechargeable, Light-weight design **Sold for research purposes only and not for human use**
Catalog No. NC9010016 Supplier Braintree Scientific Supplier No. MD1200120V
Ideal Micro-Drill Designed for research applications that require surgical burrs and trephines. Comes with five burrs: .006mm, .008mm, .010mm, 1.012mm, 1.016mm. Rechargeable, Light-weight design **Sold for research purposes only and not for human use**