Learn More
Roboz Surgical IDEAL MICRO-DRILL
SDP
Catalog No. NC9819481
Encompass
The Ideal Micro-Drill is designed for research applications that require surgical burs and trephines. The drill is constructed of light-weight aluminum alloy for balance and control and is powered by a rechargeable 6-volt nickel cadmium batter
Catalog No. NC9819481 Supplier Roboz Surgical Supplier No. RS6300A
The Ideal Micro-Drill is designed for research applications that require surgical burs and trephines. The drill is constructed of light-weight aluminum alloy for balance and control and is powered by a rechargeable 6-volt nickel cadmium batter