Learn More
Enterprise Technology Solutions Ideal Shredders Shredder Oil, High-Cling, f/2245/3104/3804, 1 Pint, Clear
SDP
Catalog No. 5000740557
Encompass_Preferred
Special High-Cling Shredder Oil lubricates your shredder blades so it runs smoothly and effectively. This nontoxic, high-cling, nondetergent oil formula is made exclusively for Ideal brand shredders. Oil shredder blades for every 30 minutes of operation for cross-cut shredders and for every hour of operation for strip-cut models. Use oil in Ideal 2245, 2265, 2270, 2360, 2404, 2445, 2465, 2503, 3104 and 3804 shredders without automatic oilers.
Catalog No. 50-007-40557 Supplier Enterprise Technology Solutions Supplier No. ISRIDEACCED214H
Special High-Cling Shredder Oil lubricates your shredder blades so it runs smoothly and effectively. This nontoxic, high-cling, nondetergent oil formula is made exclusively for Ideal brand shredders. Oil shredder blades for every 30 minutes of operation for cross-cut shredders and for every hour of operation for strip-cut models. Use oil in Ideal 2245, 2265, 2270, 2360, 2404, 2445, 2465, 2503, 3104 and 3804 shredders without automatic oilers.