Learn More
Enzo Life Sciences Idebenone (100mg). CAS: 58186-27-9
SDP
Catalog No. 502010837
Encompass_Preferred
Idebenone is a quinone antioxidant chemically related to ubiquinone. It is a free radical scavenger that protects mitochondrial membranes from lipid peroxidation. It improves overall brain function in various animal models and protective effect on astrocytes is due in part via increased NGF production. Alternative name: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione. Purity: ≥98% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-0837 Supplier Enzo Life Sciences Supplier No. BMLFR1140100
Idebenone is a quinone antioxidant chemically related to ubiquinone. It is a free radical scavenger that protects mitochondrial membranes from lipid peroxidation. It improves overall brain function in various animal models and protective effect on astrocytes is due in part via increased NGF production. Alternative name: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione. Purity: ≥98% (HPLC). Long Term Storage: Ambient.