Learn More
Enzo Life Sciences Idebenone (20mg). CAS: 58186-27-9
SDP
Supplier:  Enzo Life Sciences BMLFR1140020
Encompass_Preferred
Idebenone is a quinone antioxidant chemically related to ubiquinone. It is a free radical scavenger that protects mitochondrial membranes from lipid peroxidation. It improves overall brain function in various animal models and protective effect on astrocytes is due in part via increased NGF production. Alternative name: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione. Purity: ≥98% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-0836