Learn More
Abcam Idebenone, antio x idant and neuroprotective agent, 100MG
Catalog No. 502706224
Encompass_Preferred
MW 338.4 Da, Purity >98%. Potent antioxidant and neuroprotective agent. Coenzyme Q analogue. Active in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6224 Supplier Abcam Supplier No. AB142522100MG
MW 338.4 Da, Purity >98%. Potent antioxidant and neuroprotective agent. Coenzyme Q analogue. Active in vivo..
The product is subject to the following: Abcam Restricted Use Statement