Learn More
Abcam Idebenone, antio x idant and neuroprotective agent, 25MG
Catalog No. 502706222
Encompass_Preferred
MW 338.4 Da, Purity >98%. Potent antioxidant and neuroprotective agent. Coenzyme Q analogue. Active in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6222 Supplier Abcam Supplier No. AB14252225MG
MW 338.4 Da, Purity >98%. Potent antioxidant and neuroprotective agent. Coenzyme Q analogue. Active in vivo..
The product is subject to the following: Abcam Restricted Use Statement