Learn More
Medchemexpress LLC HY-13026 50mg Medchemexpress, Idelalisib CAS:870281-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302650MG
Encompass_Preferred
Medchemexpress, HY-13026 50mg Idelalisib CAS:870281-82-6 Idelalisib (CAL-101; GS-1101) is a highly selective and orally bioavailable p110δ inhibitor with an IC50 of 2.5 nM, showing 40- to 300-fold selectivity for p110δ over other PI3K class I enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8947