Learn More
Medchemexpress LLC Idelalisib-d5 | 1830330-31-8 | 98.4% | 1 MG
SDP
Catalog No. 5000340461
Encompass_Preferred
Idelalisib-d5 is a deuterium-labeled Idelalisib. It is a highly selective and orally bioavailable p110δ inhibitor.
  • Serves as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS techniques
Catalog No. 50-003-40461 Supplier Medchemexpress LLC Supplier No. HY13026S1MG
Idelalisib-d5 is a deuterium-labeled Idelalisib. It is a highly selective and orally bioavailable p110δ inhibitor.
  • Serves as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS techniques