Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jaece Industries Identi-Marker Pens

Catalog No. p-7078254
Change view
Click to view available options
Color:
Black
Blue
Green
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-112 Blue
14-127-111 Black
14-127-114 Red
14-127-113 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-127-112 Supplier Jaece Industries Supplier No. IMF200
This item is not returnable. View return policy

Writes on foam, plastic, glass and metal. Jaece Identi-Marker Pens are ideal for use with Identi-Plugs™ and other Jaece products.

  • Ideal for use with Identi-Plugs™ and other Jaece products
  • Writes on cold or wet surfaces
  • Marks on plastic, glass and metal
  • Permanent, quick-drying ink
  • Extra-fine, durable plastic tip
  • Ideal for general laboratory, hospital or industrial use

Specifications

Product Type Identi-Marker Pen
Color Blue
For Use With (Application) General laboratory, hospital or industrial use
For Use With (Equipment) Identi-Plugs™ and other Jaece products
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.