Learn More
Key Scientific Products IDENTICULT AE STRIPS (W/PEP), 25 per Pack
SDP
Catalog No. NC0335482
Encompass_Preferred
This test is designed to help detect and identify Strep pyogenes and Enterococci plus members of the Enterobacteriaceae from clinical isolates. IDENTICULT AE uses a chromogenic substrate, L-pyroglutamylpeptide hydrolase (PYR) which produces a red to pink color upon addition of the reagent. Reagent # 2 is cinnamaldehyde, aka Spot Indole Reagent. Thus, this test gives TWO results which can also be used in the rapid identification of E. Coli from positive urine cultures.
Catalog No. NC0335482 Supplier Key Scientific Products Supplier No. KF4080
This test is designed to help detect and identify Strep pyogenes and Enterococci plus members of the Enterobacteriaceae from clinical isolates. IDENTICULT AE uses a chromogenic substrate, L-pyroglutamylpeptide hydrolase (PYR) which produces a red to pink color upon addition of the reagent. Reagent # 2 is cinnamaldehyde, aka Spot Indole Reagent. Thus, this test gives TWO results which can also be used in the rapid identification of E. Coli from positive urine cultures.