Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Identification and Characterization of Bacteria Kit
SDPSCI_ED

Catalog No. S27555 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27555 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27555 Supplier Edvotek™ Supplier No. 160
Add to Cart
Add to Cart

Gram staining is a quick, effective, and medically relevant identification test that is essential for bacterial classification. Students will use this staining technique to examine the size, shape, arrangement, and gram status (+/-) of Escherichia coli, Bacillus subtilis, and Micrococcus luteus.

  • Storage: Some Components Require Refrigerator Storage Upon Receipt
  • Completion Time: 90 minutes
  • For 8 Groups

Specifications

Grade 9 to Undergraduate
Age 14 to 20
Includes Instructions, BactoBeads™, ReadyPour™ Luria Broth Agar, Large Petri Plates, 10 mL Sterile Pipette, Inoculating Loops, Toothpicks, Transfer Pipettes, Microscope Slides, Gram's Crystal Violet, Gram's Iodine, Gram's Safranin, Sterile Water, Microcentrifuge Tubes
Requires Microscopes, Cover Slips, Microwave, Pipette Pump, Timers, Slide Holders, Bunsen Burner or Alcohol Lamp, Lab Marker, Beakers, Small Staining Tray, Paper Towels, 95% Ethanol, Distilled Water, Gloves
Time Range 90 min.
Content And Storage Some Components Require Refrigerator Storage Upon Receipt
Class Size 8 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.