Learn More
EDVOTEK™ Identifying Red Snapper Using Genetic Analysis
SDPSCI_ED
Catalog No. S000289 Shop All EDVOTEK Products
Change view
Click to view available options
Product Type:
DNA Bioinformatics Experiment
Catalog No. Product Type
S000289 DNA Bioinformatics Experiment
1 options
Catalog No. S000289 Supplier EDVOTEK™ Supplier No. 341
Add to Cart
Add to Cart
Extract DNA from simulated fish samples using PCR
  • Learn how to extract DNA from simulated fish samples and amplify specific gene regions using PCR
  • Understand how conserved gene regions can be used to identify species and study genetic diversity
  • Explore DNA barcoding, a critical tool in species identification and conservation biology
  • In this experiment, students will explore the genetic diversity of fish species in a simulated DNA barcoding experiment
  • Students will extract DNA from simulated fish samples and use PCR to amplify three gene regions that are conserved between species
Specifications
Class Size 24 Students (6 Groups)
Product Type DNA Bioinformatics Experiment