Learn More
Cole-Parmer Idex 1569 Chromatography Tubing, Orange PEEK, 1/32" OD x 0.020" ID x 5 ft L; 1/EA
Catalog No. NC1764592 Shop All Cole-Parmer Products
Encompass
Applications: Chromatography Characteristics: Unlike stainless steel, PEEK (polyetheretherketone) tubing is flexible and easy to cut. It provides excellent chemical resistance to both organic and inorganic solvents. Tubing withstands pressures to 5000 psi (3000 psi for model 02006-30). PEEK tubing has a very smooth internal surface, which causes less turbulence than metal tubing and improves resolution. Pack of 5-ft. Certification: None Maximum temperature: 212°F (100°C) Sterilize: With autoclave, ethylene oxide, or gamma irradiation
Catalog No. NC1764592 Supplier Cole-Parmer Supplier No. 0200611
Applications: Chromatography Characteristics: Unlike stainless steel, PEEK (polyetheretherketone) tubing is flexible and easy to cut. It provides excellent chemical resistance to both organic and inorganic solvents. Tubing withstands pressures to 5000 psi (3000 psi for model 02006-30). PEEK tubing has a very smooth internal surface, which causes less turbulence than metal tubing and improves resolution. Pack of 5-ft. Certification: None Maximum temperature: 212°F (100°C) Sterilize: With autoclave, ethylene oxide, or gamma irradiation