Learn More
Cole-Parmer Idex P-754 Tubing Cap, Yellow ETFE, 10-32 Coned; 1/Ea
Supplier:  Cole-Parmer 0201848
Encompass
Caps are made of ETFE for increased strength and low gas permeability.
Catalog No. NC1713340