Learn More
Cole-Parmer Idex Extender Tool for Standard Head Nuts, Aluminum; 1/EA
Supplier:  Cole-Parmer 0202549
Encompass
Tighten most of the knurled nuts in hard-to-reach places for leak-tight connections using extender tools. Simply match up the head style to the tool. Use extender tools to torque driver 02022-80 for tightening to a specific amount of tightening torque or as stand-alone units.
Catalog No. NC2229485