Learn More
Trajan Scientific Idex One-Piece Fingertight, Natural, 10-32 Coned, for 1/16"" OD - 10 Pack
Catalog No. 501871388
Encompass
One-Piece Fingertight, Idex Natural, 10-32 Coned, for 1/16" OD - 10 Pack
Catalog No. 50-187-1388 Supplier Trajan Scientific Supplier No. ID.F120X
One-Piece Fingertight, Idex Natural, 10-32 Coned, for 1/16" OD - 10 Pack