Learn More
Cole-Parmer Idex Flangeless Ferrule, Blue ETFE, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/Ea
Catalog No. NC2186227 Shop All Cole-Parmer Products
Encompass
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
This item is not returnable. View return policy
Catalog No. NC2186227 Supplier Cole-Parmer Supplier No. 1202967
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.