Learn More
Trajan Scientific IDEX Flangeless Nut Short, PEEK 1/4-28 Flat-Bottom, for 1/8"" OD, 10/pk
Catalog No. 501871423
Encompass
IDEX Flangeless Nut Short, PEEK 1/4-28 Flat-Bottom, for 1/8" OD, 10/pk
Catalog No. 50-187-1423 Supplier Trajan Scientific Supplier No. ID.P335X
IDEX Flangeless Nut Short, PEEK 1/4-28 Flat-Bottom, for 1/8" OD, 10/pk