Learn More
IDEX Health & Science Upchurch Semi-Prep Stainless Steel Frit 0.5µm .062" x .065" x .200"
Catalog No. NC2016325
Encompass_Preferred
This item has a minimum order quantity of 25 per supplier requirements. Semi-Prep Stainless Steel Frit 0.5µm .062" x .065" x .200"
Catalog No. NC2016325 Supplier IDEX Health & Science Upchurch Supplier No. A418
This item has a minimum order quantity of 25 per supplier requirements. Semi-Prep Stainless Steel Frit 0.5µm .062" x .065" x .200"