Learn More
Cole-Parmer IDEX P-232X SUPER FLANGELESS
Catalog No. NC1991122 Shop All Cole-Parmer Products
Encompass
Idex P-232X Super Flangeless Nut, Headless, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Catalog No. NC1991122 Supplier Cole-Parmer Supplier No. 0201371
Idex P-232X Super Flangeless Nut, Headless, Short, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK