Learn More
Cole-Parmer Idex Flangeless Male Nut, Short Head, Black PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
Catalog No. NC2045928 Shop All Cole-Parmer Products
Encompass
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Ferrules are available in various materials to suit your application.
Catalog No. NC2045928 Supplier Cole-Parmer Supplier No. 0202039
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Ferrules are available in various materials to suit your application.