Learn More
Cole-Parmer Idex Super Flangeless™ Nut, Standard Knurl, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Catalog No. NC2008815 Shop All Cole-Parmer Products
Encompass
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Catalog No. NC2008815 Supplier Cole-Parmer Supplier No. 0201386
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.