Learn More
Cole-Parmer Idex Threaded Luer Adapter, Natural/Black PEEK, 0.050" Bore, Male Luer Lock x Female 10-32 Coned
Catalog No. NC2045929 Shop All Cole-Parmer Products
Encompass
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Catalog No. NC2045929 Supplier Cole-Parmer Supplier No. 0201413
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.