Learn More
Cole-Parmer Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA
Catalog No. NC2081349 Shop All Cole-Parmer Products
Encompass
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Catalog No. NC2081349 Supplier Cole-Parmer Supplier No. 0202358
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.