Learn More
Cole-Parmer Idex Peristaltic Tubing Adapter, PEEK/ETFE, 0.030" Bore, up to 0.18" OD x 0.048" to 0.11" ID
Catalog No. NC2023066 Shop All Cole-Parmer Products
Encompass
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.
Catalog No. NC2023066 Supplier Cole-Parmer Supplier No. 0201439
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.