Learn More
Trajan Scientific Idex PEEK Tubing, blue, 1/16"" OD X 010 mm, 7000 psi, 100 feet
Catalog No. 501871268
Encompass
PEEK Tubing, Idex, blue, 1/16" OD X 010 mm, 7000 psi, 100 feet
Catalog No. 50-187-1268 Supplier Trajan Scientific Supplier No. ID.1531BXL
PEEK Tubing, Idex, blue, 1/16" OD X 010 mm, 7000 psi, 100 feet