Learn More
Cole-Parmer IDEX U-190 CHROMATOGRAPHY TUBI
Supplier:  Cole-Parmer 0201172
Encompass
Idex U-190 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 100 ft L, Natural; 1/EA
Catalog No. NC2134021