Learn More
Trajan Scientific Idex UltraLife Switching Valve, for MX Series II, 2 Position, 10 Port, 1/16"", 0.20mm, Max Pressure: 15,000 psi
Catalog No. 501871402
Encompass
UltraLife Switching Valve, Idex/Rheodyne, for MX Series II, 2 Position, 10 Port, 1/16", 0.20mm, Max Pressure: 15,000 psi
Catalog No. 50-187-1402 Supplier Trajan Scientific Supplier No. ID.MXT715102
UltraLife Switching Valve, Idex/Rheodyne, for MX Series II, 2 Position, 10 Port, 1/16", 0.20mm, Max Pressure: 15,000 psi