Learn More
Medchemexpress LLC HY-111387 5mg Medchemexpress, IDF-11774 CAS:1429054-28-3 Purity:>98%
SDP
Catalog No. 501968851
Encompass_Preferred
Medchemexpress, HY-111387 5mg IDF-11774 CAS:1429054-28-3 IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8851 Supplier Medchemexpress LLC Supplier No. HY1113875MG
Medchemexpress, HY-111387 5mg IDF-11774 CAS:1429054-28-3 IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.