Learn More
Selleck Chemical LLC IDF-11774 5mg 1429054-28-3
SDP
Supplier:  Selleck Chemical LLC S87715MG
Encompass_Preferred
IDF-11774 is a hypoxia-inducible factor-1 (HIF-1) inhibitor. It reduces the HRE-luciferase activity of HIF-1? (IC50 = 3.65 ?M) and blocks HIF-1? accumulation under hypoxia in HCT116 human colon cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8300