Learn More
Aviva Systems Biology Corp IDH1 Antibody - C-terminal region (ARP54787_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IDH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human IDH1. Alternate names: IDH, IDP, PICD, IDCD, IDPCApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-53744