Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human isocitrate dehydrogenase [NADP] cytoplasmic (IDH1), His-tagged | 96.7% | 100UG
SDP

Catalog No. 5000176040
Encompass_Preferred

Recombinant full-length human isocitrate dehydrogenase 1 (IDH1), expressed in HEK293 cells and purified with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in a stabilized buffer for biochemical and structural research, available in multiple sizes and shipped frozen on dry ice.

  • Expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification.
  • Full length protein (M1-L414), approx. 48 kDa.
  • Supplied in stabilized buffer (Tris-HCl, NaCl, Brij-35, glycerol, DTT).
  • Purity greater than 95% (96.7% by reducing SDS-PAGE).
  • Store at -80°C; ship on dry ice; avoid repeated freeze-thaw.
  • Available sizes include 10 μg, 50 μg, and 100 μg.

Catalog No. 50-001-76040 Supplier Medchemexpress LLC Supplier No. HYP70214100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human isocitrate dehydrogenase 1 (IDH1), expressed in HEK293 cells and purified with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in a stabilized buffer for biochemical and structural research, available in multiple sizes and shipped frozen on dry ice.

  • Expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification.
  • Full length protein (M1-L414), approx. 48 kDa.
  • Supplied in stabilized buffer (Tris-HCl, NaCl, Brij-35, glycerol, DTT).
  • Purity greater than 95% (96.7% by reducing SDS-PAGE).
  • Store at -80°C; ship on dry ice; avoid repeated freeze-thaw.
  • Available sizes include 10 μg, 50 μg, and 100 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.