Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isocitrate dehydrogenase 1 protein, human | 96.7% | 100UG
SDP

Catalog No. 5000176792
Encompass_Preferred

Recombinant human isocitrate dehydrogenase 1 (IDH1) expressed in HEK293 cells, supplied as a filtered solution with a C-terminal 6xHis tag. The protein is provided at approximately 0.87 mg/mL in 20 mM Tris-HCl, 150 mM NaCl, 0.05% Brij-35, 10% glycerol, 1 mM DTT, pH 8.0, and is intended for biochemical assays, structural studies, and antibody generation.

  • Recombinant human IDH1 with mammalian post-translational modifications.
  • C-terminal 6xHis tag for purification and detection.
  • Supplied as a 0.22 μm filtered solution at approximately 0.87 mg/mL.
  • Purity greater than 95% (96.7%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 48 kDa.
  • Formulated with glycerol and DTT for stability; avoid repeated freeze-thaw cycles.

Catalog No. 50-001-76792 Supplier Medchemexpress LLC Supplier No. HYP70214A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human isocitrate dehydrogenase 1 (IDH1) expressed in HEK293 cells, supplied as a filtered solution with a C-terminal 6xHis tag. The protein is provided at approximately 0.87 mg/mL in 20 mM Tris-HCl, 150 mM NaCl, 0.05% Brij-35, 10% glycerol, 1 mM DTT, pH 8.0, and is intended for biochemical assays, structural studies, and antibody generation.

  • Recombinant human IDH1 with mammalian post-translational modifications.
  • C-terminal 6xHis tag for purification and detection.
  • Supplied as a 0.22 μm filtered solution at approximately 0.87 mg/mL.
  • Purity greater than 95% (96.7%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 48 kDa.
  • Formulated with glycerol and DTT for stability; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.