Learn More
Medchemexpress LLC IDH1 protein, human (HEK293, His, solution) | 96.7% | 50UG
SDP
Catalog No. 5000176047
Encompass_Preferred
Recombinant human isocitrate dehydrogenase 1 (IDH1) produced in HEK293 cells and supplied as a filtered, buffered solution with a C-terminal 6xHis tag. The protein is intended for biochemical assays, structural studies, and assay or antibody development; batch-specific COA provides purity, endotoxin, and handling details.
Catalog No. 50-001-76047 Supplier Medchemexpress LLC Supplier No. HYP70214A50UG
Recombinant human isocitrate dehydrogenase 1 (IDH1) produced in HEK293 cells and supplied as a filtered, buffered solution with a C-terminal 6xHis tag. The protein is intended for biochemical assays, structural studies, and assay or antibody development; batch-specific COA provides purity, endotoxin, and handling details.