Learn More
Medchemexpress LLC IDH1 inhibitor 1 | 2234285-81-3 | 100.0% | 450.39 g/mol | C20H18F4N6O2 | 5 MG
SDP
Catalog No. 5000262693
Encompass_Preferred
IDH1 inhibitor 1 is a small-molecule research compound that selectively inhibits mutant isocitrate dehydrogenase 1 (IDH1), reducing the oncometabolite 2-hydroxyglutarate and showing activity in preclinical cancer models. It is reported to be orally bioavailable and brain-penetrant, and is provided for laboratory research use.
  • Selective inhibitor of mutant IDH1 enzymes.
  • Reduces 2-hydroxyglutarate (2-HG) production in cell and animal models.
  • Orally bioavailable with reported brain penetration.
  • High reported purity suitable for research applications.
  • Available in small-mass packages for preclinical studies.
Catalog No. 50-002-62693 Supplier Medchemexpress LLC Supplier No. HY1126015MG
IDH1 inhibitor 1 is a small-molecule research compound that selectively inhibits mutant isocitrate dehydrogenase 1 (IDH1), reducing the oncometabolite 2-hydroxyglutarate and showing activity in preclinical cancer models. It is reported to be orally bioavailable and brain-penetrant, and is provided for laboratory research use.
  • Selective inhibitor of mutant IDH1 enzymes.
  • Reduces 2-hydroxyglutarate (2-HG) production in cell and animal models.
  • Orally bioavailable with reported brain penetration.
  • High reported purity suitable for research applications.
  • Available in small-mass packages for preclinical studies.