Learn More
Medchemexpress LLC IDH1 Inhibitor 7 | 2135309-56-5 | C22H24F3N7O | 5 MG
SDP
Catalog No. 5000375537
Encompass_Preferred
IDH1 Inhibitor 7 (Compound 88) is a potent IDH1 inhibitor with an IC50 of less than 100 nM. It is a white to off-white solid with a molecular weight of 459.47 and is soluble in DMSO at 100 mg/mL. This compound is intended for research use only.
  • IC50 of less than 100 nM against IDH1
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-75537 Supplier Medchemexpress LLC Supplier No. HY1502385MG
IDH1 Inhibitor 7 (Compound 88) is a potent IDH1 inhibitor with an IC50 of less than 100 nM. It is a white to off-white solid with a molecular weight of 459.47 and is soluble in DMSO at 100 mg/mL. This compound is intended for research use only.
  • IC50 of less than 100 nM against IDH1
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL