Learn More
Medchemexpress LLC IDH1 inhibitor 7-d2 | 2135309-51-0 | C₂₂H₂₂D₂F₃N₃O | 1 MG
SDP
Catalog No. 5000374772
Encompass_Preferred
IDH1 Inhibitor 7-d2 is a deuterium labeled IDH1 Inhibitor 7, which functions as an IDH1 inhibitor with an IC₅₀ of less than 100 nM. This compound incorporates stable heavy isotopes, which can be used as tracers during drug development for quantitation and may influence the pharmacokinetic and metabolic profiles of drugs.
  • IDH1 inhibitor with an IC₅₀ of less than 100 nM
  • Incorporates stable heavy isotopes for potential use as tracers
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74772 Supplier Medchemexpress LLC Supplier No. HY150238S1MG
IDH1 Inhibitor 7-d2 is a deuterium labeled IDH1 Inhibitor 7, which functions as an IDH1 inhibitor with an IC₅₀ of less than 100 nM. This compound incorporates stable heavy isotopes, which can be used as tracers during drug development for quantitation and may influence the pharmacokinetic and metabolic profiles of drugs.
  • IDH1 inhibitor with an IC₅₀ of less than 100 nM
  • Incorporates stable heavy isotopes for potential use as tracers
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs