Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IDH1 inhibitor 7-d2 | 2135309-51-0 | MFCD00047640 | 99.9% | 461.48 | C22H22D2F3N7O | 10 MG
SDP

Catalog No. 5000286127
Encompass_Preferred

IDH1 inhibitor 7-d2 is the deuterium-labeled analogue of IDH1 inhibitor 7 supplied as a high-purity solid for research applications. It is intended for use as a stable isotope tracer or internal standard in biochemical and analytical studies of IDH1 inhibition, and is provided with specified storage recommendations to maintain compound integrity.

  • Deuterium-labeled tracer for quantification and metabolic studies.
  • High purity suitable for analytical applications.
  • Solid (powder) form for convenient handling and storage.
  • Well-defined molecular properties: formula C22H22D2F3N7O, molecular weight 461.48.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-86127 Supplier Medchemexpress LLC Supplier No. HY150238S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IDH1 inhibitor 7-d2 is the deuterium-labeled analogue of IDH1 inhibitor 7 supplied as a high-purity solid for research applications. It is intended for use as a stable isotope tracer or internal standard in biochemical and analytical studies of IDH1 inhibition, and is provided with specified storage recommendations to maintain compound integrity.

  • Deuterium-labeled tracer for quantification and metabolic studies.
  • High purity suitable for analytical applications.
  • Solid (powder) form for convenient handling and storage.
  • Well-defined molecular properties: formula C22H22D2F3N7O, molecular weight 461.48.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.