Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IDH1 Inhibitor 8 | 1448346-63-1 | 99.0% | 582.96 | 1 MG
SDP

Catalog No. 5000403645
Encompass_Preferred

IDH1 Inhibitor 8 (91) is an isocitrate dehydrogenase 1 (IDH1) inhibitor. It is intended for use in cancer research. This product is for research use only and is not sold to patients.

  • Target: Isocitrate dehydrogenase 1 (IDH1)
  • Application: Cancer research
  • Appearance: Solid, white to off-white
  • Solubility: Soluble in DMSO (100 mg/mL, 171.54 mM, with ultrasonic; use newly opened DMSO)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Catalog No. 50-004-03645 Supplier Medchemexpress LLC Supplier No. HY416481MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IDH1 Inhibitor 8 (91) is an isocitrate dehydrogenase 1 (IDH1) inhibitor. It is intended for use in cancer research. This product is for research use only and is not sold to patients.

  • Target: Isocitrate dehydrogenase 1 (IDH1)
  • Application: Cancer research
  • Appearance: Solid, white to off-white
  • Solubility: Soluble in DMSO (100 mg/mL, 171.54 mM, with ultrasonic; use newly opened DMSO)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.