Learn More
Cayman Chemical IDH1R132H mutnthumnre 100ug
SDP
Catalog No. 502954406
Encompass_Preferred
Source: Active recombinant C-terminal His-tagged protein expressed in E. coli • MW: 47.9 kDa
Catalog No. 50-295-4406 Supplier Cayman Chemical Supplier No. 14132100 UG
Source: Active recombinant C-terminal His-tagged protein expressed in E. coli • MW: 47.9 kDa